Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich p-Divinylbenzene

Catalog No. 5355835ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-558-35ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 53-558-35ML Supplier Sigma Aldrich Supplier No. 5355835ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

p-Divinylbenzene(DVB) is a cross-linking agent, which is used in a variety of polymeric processes such as copolymerization of styrene to improve the thermo-mechanical properties of the composite.DVB can be cross-linked with styrene/vinylbenzene chloride and incorporated with multi-walled carbon nanotubes (MWCNTs) to form new microporous nanocomposites for water remediation. It can also be used in combination with polyethylene glycol diacrylate to form linkages with polyethylene oxide (PEO), which can be used as an electrolytic material for lithium batteries.

Specifications

CAS 105-06-6
Density 0.914 g/mL (at 25°C)
Molecular Formula C10H10
Refractive Index n20/D 1.5470 (literature)
Linear Formula C10H10
Quantity 5 mL
MDL Number MFCD00008622
Synonym 1,4-Diethenylbenzene; 1,4-Divinylbenzene
Molecular Weight (g/mol) 130.19
Percent Purity 85%
Biological Activity Respiratory system
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.