Learn More
Mopec P-DMAC Paper
SDP
Catalog No. 22446318 Shop All Mopec Products
Change view
Click to view available options
For Use With (Application):
Latent Fingerprint Development
Catalog No. For Use With (Application)
22-446-318 Latent Fingerprint Development
1 options
Catalog No. 22-446-318 Supplier Mopec Supplier No. FS470
Add to Cart
Add to Cart
Designed to develop latent fingerprints on porous, heat-sensitive surfaces such as thermal paper and receipts, the treated paper reacts with fingerprint residue to produce fluorescent ridge detail that can be viewed under an appropriate alternate light source for examination.
Performance
  • 6 month shelf life
  • Best results usually visualized between 475nm and 515nm with an orange barrier filter or glasses
  • To extend longevity of this product, always store in the provided amber sleeve
Specifications
For Use With (Application) Latent Fingerprint Development