Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Dodecylaniline (Technical), MP Biomedicals

Catalog No. ICN206249 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN206249 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN206249 Supplier MP Biomedicals, Inc Supplier No. 0520624905

Chemical Identifiers

CAS 104-42-7
Molecular Formula C18H31N
Molecular Weight (g/mol) 261.453
InChI Key KLPPPIIIEMUEGP-UHFFFAOYSA-N
Synonym p-dodecylaniline, benzenamine, 4-dodecyl, p-dodecyl aniline, 4-dodecylphenylamine, 4-n-dodecylamine, p-n-dodecylaniline, paradodecyl aniline, acmc-20amjq, 4-dodecylaniline, ksc180i8p
PubChem CID 7701
IUPAC Name 4-dodecylaniline
SMILES CCCCCCCCCCCCC1=CC=C(C=C1)N

Specifications

Quantity 5 g
Chemical Name or Material p-Dodecylaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.