Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC P-ethylhydratropic acid | 3585-52-2 | 99.5% | 178.23 g·mol-1 | C11H14O2 | 10 MG
SDP

Catalog No. 5000208343
Encompass_Preferred

p-Ethylhydratropic acid is an identified impurity of ibuprofen supplied as an analytical reference standard for impurity profiling and method development.

  • impurity of ibuprofen used for analytical reference and profiling.
  • cas number 3585-52-2.
  • molecular formula c11h14o2 and molecular weight 178.23 g·mol-1.
  • high purity by hplc (≈99.5%).
  • common package sizes: 5 mg, 10 mg.
  • storage: store at room temperature (3 years); in solvent -80°C (2 years), -20°C (1 year).
  • intended for research use only; not for human use.

Catalog No. 50-002-08343 Supplier Medchemexpress LLC Supplier No. HYW08719410MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

p-Ethylhydratropic acid is an identified impurity of ibuprofen supplied as an analytical reference standard for impurity profiling and method development.

  • impurity of ibuprofen used for analytical reference and profiling.
  • cas number 3585-52-2.
  • molecular formula c11h14o2 and molecular weight 178.23 g·mol-1.
  • high purity by hplc (≈99.5%).
  • common package sizes: 5 mg, 10 mg.
  • storage: store at room temperature (3 years); in solvent -80°C (2 years), -20°C (1 year).
  • intended for research use only; not for human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.