Learn More
Medchemexpress LLC p-Ethylhydratropic acid | 3585-52-2 | 99.6% | 178.23 g·mol^-1 | C11H14O2 | 5 MG
SDP
Catalog No. 5000208344
Encompass_Preferred
p-Ethylhydratropic acid is a small-molecule impurity of ibuprofen used as an analytical reference for impurity profiling and method development. The compound is a solid powder with chemical formula C11H14O2, molecular weight 178.23 g·mol^-1, and CAS number 3585-52-2. Purity is reported at 99.6%, and the substance is typically supplied in milligram quantities for laboratory analysis.
  • high-purity analytical standard for impurity identification.
  • supports analytical method development and validation.
  • suitable for impurity profiling and comparability studies.
  • provided in low-mass vials for laboratory use.
  • clear chemical identifiers for traceability (CAS and molecular formula).
Catalog No. 50-002-08344 Supplier Medchemexpress LLC Supplier No. HYW0871945MG
p-Ethylhydratropic acid is a small-molecule impurity of ibuprofen used as an analytical reference for impurity profiling and method development. The compound is a solid powder with chemical formula C11H14O2, molecular weight 178.23 g·mol^-1, and CAS number 3585-52-2. Purity is reported at 99.6%, and the substance is typically supplied in milligram quantities for laboratory analysis.
  • high-purity analytical standard for impurity identification.
  • supports analytical method development and validation.
  • suitable for impurity profiling and comparability studies.
  • provided in low-mass vials for laboratory use.
  • clear chemical identifiers for traceability (CAS and molecular formula).