Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Ethyltoluene 98%, Thermo Scientific™

Catalog No. AC119010100
Change view
Click to view available options
Quantity:
10 mL
50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC119010100 10 mL
AC119010500 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC119010100 Supplier Thermo Scientific Chemicals Supplier No. 119010100

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 622-96-8
Molecular Formula C9H12
Molecular Weight (g/mol) 120.195
MDL Number MFCD00009263
InChI Key JRLPEMVDPFPYPJ-UHFFFAOYSA-N
Synonym 4-ethyltoluene, p-ethyltoluene, benzene, 1-ethyl-4-methyl, 4-methylethylbenzene, toluene, p-ethyl, p-ethylmethylbenzene, p-methylethylbenzene, 1-methyl-4-ethylbenzene, 1-ethyl-4-methyl-benzene, para-ethyltoluene
PubChem CID 12160
IUPAC Name 1-ethyl-4-methylbenzene
SMILES CCC1=CC=C(C=C1)C

Specifications

Color Colorless to Yellow
Density 0.86g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Refractive Index 1.4940 to 1.4960 (20°C, 589nm)
Linear Formula C2H5C6H4CH3
Quantity 10 mL
Beilstein 05, 397
Formula Weight 120.19
Percent Purity ≥97.5%
Physical Form Liquid
Chemical Name or Material p-Ethyltoluene, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
May be fatal if swallowed and enters airways.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 210-761-2
RTECSNumber XT2550000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.