Learn More
TARGETMOL CHEMICALS INC P-ETHYNYLPHENYLALANINE 10MG
SDP
Catalog No. 502581760
Encompass_Preferred
Also available in 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. p-Ethynylphenylalanine (4-Ethynyl-L-phenylalanine) a tryptophan hydroxylase (TPH) inhibitor is competitive effective selective and reversible with a Ki of 32.6 uM. purity: 98%
Catalog No. 50-258-1760 Supplier TARGETMOL CHEMICALS INC Supplier No. T1014210MG
Also available in 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. p-Ethynylphenylalanine (4-Ethynyl-L-phenylalanine) a tryptophan hydroxylase (TPH) inhibitor is competitive effective selective and reversible with a Ki of 32.6 uM. purity: 98%