Learn More
CPC Scientific H-Pro-Phe-Thr-Arg-Asn-Tyr-Tyr-Val-Arg-Ala-Val-Leu-His-Leu-OH (trifluoroacetate salt) 5MG
SDP
Catalog No. 502108773
Encompass
ONE-LETTER SEQUENCE: PFTRNYYVRAVLHLMOLECULAR FORMULA: C83H125N23O19MOLECULAR WEIGHT:1749.1STORAGE CONDITIONS: -20 5CRESEARCH AREA: CancerREFERENCES: M.-Y.Park et al., Biochem. Biophys. Res. Commun. , 204, 924 (1994)
Catalog No. 50-210-8773 Supplier CPC Scientific Supplier No. AOXP002B
ONE-LETTER SEQUENCE: PFTRNYYVRAVLHLMOLECULAR FORMULA: C83H125N23O19MOLECULAR WEIGHT:1749.1STORAGE CONDITIONS: -20 5CRESEARCH AREA: CancerREFERENCES: M.-Y.Park et al., Biochem. Biophys. Res. Commun. , 204, 924 (1994)