Learn More
Sino Biological Plasmodium falciparum (strain 3D7) CSP Protein (His & GST Tag)
Supplier:  Sino Biological 69089V53E100
Encompass_Preferred
A DNA sequence encoding the P. falciparum CSP (Q7K740) (Leu19-Pro104) was fused with the N-terminal polyhistidine tag followed by a GST tag.
Catalog No. 50-255-2905