Learn More
Sino Biological Anti-PfLDH Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 40303RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant P. falciparum PfLDH / L-lactate dehydrogenase (Catalog#40303-N08E; XP_001349989.1; Met1-Ala316). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-3728