Learn More
BUCHI P+G Coated Glass Column, 49 mm I.D.
SDP
Catalog No. 05405445 Shop All BUCHI Products
Change view
Click to view available options
Glass
Catalog No.
05-405-445 Glass
1 options
Catalog No. 05-405-445 Supplier BUCHI Supplier No. 019676
Add to Cart
Add to Cart
Glass column features a P+G safety coating, and an inner diameter of 49 mm.
  • P+G coated
  • Inner Diameter: 49 mm
Specifications
Material Glass
Type Flash Column
For Use With (Application) Chromatography