Learn More
BYK Gardner P-Glass Cuvette, Square, 10 x 10 mm
SDP
Catalog No. NC1766319
Encompass
For color measurement of liquids square, rectangle and cylindrical cuvettes can be used. Both precision as well as inexpensive disposable tubes are available. The precision cuvettes ensure reproducible results – even for critical solutions as clear as water. For daily QC disposable cuvettes save time and money.
This item is not returnable. View return policy
Catalog No. NC1766319 Supplier BYK Gardner Supplier No. 9508
For color measurement of liquids square, rectangle and cylindrical cuvettes can be used. Both precision as well as inexpensive disposable tubes are available. The precision cuvettes ensure reproducible results – even for critical solutions as clear as water. For daily QC disposable cuvettes save time and money.