Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC P glycoprotein antibody (YA255) | 10 UL
SDP

Catalog No. 5000442275
Encompass_Preferred

P Glycoprotein Antibody (YA255) (PBS only) is a rabbit-derived, non-conjugated IgG monoclonal antibody targeting P Glycoprotein. It is supplied in PBS, pH 7.4. P Glycoprotein is a membrane-associated protein involved in multidrug resistance as an ATP-dependent drug efflux pump for xenobiotic compounds.

  • Target specificity for P Glycoprotein.
  • Rabbit host with human reactivity.
  • Non-conjugated antibody.
  • Formulated in PBS, pH 7.4.
  • Suitable for Western Blot (WB) at 1:1000-1:50000 dilution.
  • Suitable for Immunohistochemistry-Paraffin (IHC-P) at 1:100-1:1200 dilution.
  • Store at -20°C for 1 year.
  • Avoid repeated freeze/thaw cycles.
  • Liquid appearance.
  • Shipped with blue ice.

Catalog No. 50-004-42275 Supplier Medchemexpress LLC Supplier No. HYP8077210UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

P Glycoprotein Antibody (YA255) (PBS only) is a rabbit-derived, non-conjugated IgG monoclonal antibody targeting P Glycoprotein. It is supplied in PBS, pH 7.4. P Glycoprotein is a membrane-associated protein involved in multidrug resistance as an ATP-dependent drug efflux pump for xenobiotic compounds.

  • Target specificity for P Glycoprotein.
  • Rabbit host with human reactivity.
  • Non-conjugated antibody.
  • Formulated in PBS, pH 7.4.
  • Suitable for Western Blot (WB) at 1:1000-1:50000 dilution.
  • Suitable for Immunohistochemistry-Paraffin (IHC-P) at 1:100-1:1200 dilution.
  • Store at -20°C for 1 year.
  • Avoid repeated freeze/thaw cycles.
  • Liquid appearance.
  • Shipped with blue ice.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.