Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC P Glycoprotein Antibody (YA255) | 100 UL
SDP

Catalog No. 5000442125
Encompass_Preferred

P Glycoprotein Antibody (YA255) is a rabbit-derived, non-conjugated IgG monoclonal antibody targeting P Glycoprotein, a member of the ATP-binding cassette (ABC) transporter superfamily. P Glycoprotein acts as an ATP-dependent drug efflux pump, contributing to multidrug resistance in cells and functioning as a transporter in the blood-brain barrier. It is associated with colchicine resistance and Inflammatory bowel disease 13.

  • Host: Rabbit
  • Reactivity: Human
  • Conjugation: Non-conjugated
  • SwissProt ID: P08183
  • Research field: Signal transduction
  • Predicted molecular weight: 141 kDa
  • Observed molecular weight: 130-180 kDa
  • Formulation: 50 mM Tris-Glycine (pH 7.4), 0.15 M NaCl, 40% Glycerol, 0.05% BSA, 0.01% Sodium azide
  • Appearance: Liquid
  • Applications: WB, IHC-P
  • Storage: Stored at -20°C for 1 year; avoid repeated freeze/thaw cycles

Catalog No. 50-004-42125 Supplier Medchemexpress LLC Supplier No. HYP80772100UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

P Glycoprotein Antibody (YA255) is a rabbit-derived, non-conjugated IgG monoclonal antibody targeting P Glycoprotein, a member of the ATP-binding cassette (ABC) transporter superfamily. P Glycoprotein acts as an ATP-dependent drug efflux pump, contributing to multidrug resistance in cells and functioning as a transporter in the blood-brain barrier. It is associated with colchicine resistance and Inflammatory bowel disease 13.

  • Host: Rabbit
  • Reactivity: Human
  • Conjugation: Non-conjugated
  • SwissProt ID: P08183
  • Research field: Signal transduction
  • Predicted molecular weight: 141 kDa
  • Observed molecular weight: 130-180 kDa
  • Formulation: 50 mM Tris-Glycine (pH 7.4), 0.15 M NaCl, 40% Glycerol, 0.05% BSA, 0.01% Sodium azide
  • Appearance: Liquid
  • Applications: WB, IHC-P
  • Storage: Stored at -20°C for 1 year; avoid repeated freeze/thaw cycles

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.