Learn More
Enzo Life Sciences P-glycoprotein monoclonal antibody (C219) (1ml)
SDP
Supplier:  Enzo Life Sciences ALX801002C100
Encompass_Preferred
Clone: C219. Isotype: IgG1. Immunogen: SDS-solubilized plasma membranes of an MDR (multidrug resistant) Chinese hamster ovary (CHO) cell line and a human cell line. Species reactivity: Human, Mouse, Rat Primate, Dog. Specificity: Recognizes an internal, highly conserved amino acid sequence (VQEALD and VQAALD) found in both protein isoforms, P-glycoprotein and MDR3 P-glycoprotein. Applications: ICC, IHC (FS), IHC (PS), IP. Formulation: Liquid. In 0.01M PBS containing 1% BSA and 0.1% sodium azide.
Catalog No. 50-200-9680