Learn More
ABclonal Technology P Glycoprotein Rabbit mAb
SDP
Catalog No. 5000011104
Encompass_Preferred
The membrane-associated protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters ABC proteins transport various molecules across extra- and intra-cellular membranes ABC genes are divided into seven distinct subfamilies (ABC1 MDR TAP MRP ALD OABP GCN20 White) This protein is a member of the MDR TAP subfamily Members of the MDR TAP subfamily are involved in multidrug resistance The protein encoded by this gene is an ATP-dependent drug efflux pump for xenobiotic compounds with broad substrate specificity It is responsible for decreased drug accumulation in multidrug-resistant cells and often mediates the development of resistance to anticancer drugs This protein also functions as a transporter in the blood-brain barrier Mutations in this gene are associated with colchicine resistance and Inflammatory bowel disease 13 Alternative splicing and the use of alternative promoters results in multiple transcript variants
Catalog No. 50-000-11104 Supplier ABclonal Technology Supplier No. A27707100UL
The membrane-associated protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters ABC proteins transport various molecules across extra- and intra-cellular membranes ABC genes are divided into seven distinct subfamilies (ABC1 MDR TAP MRP ALD OABP GCN20 White) This protein is a member of the MDR TAP subfamily Members of the MDR TAP subfamily are involved in multidrug resistance The protein encoded by this gene is an ATP-dependent drug efflux pump for xenobiotic compounds with broad substrate specificity It is responsible for decreased drug accumulation in multidrug-resistant cells and often mediates the development of resistance to anticancer drugs This protein also functions as a transporter in the blood-brain barrier Mutations in this gene are associated with colchicine resistance and Inflammatory bowel disease 13 Alternative splicing and the use of alternative promoters results in multiple transcript variants