Learn More
Medchemexpress LLC P-gp inhibitor 1 | 2050747-49-2 | 98.8% | 517.62 g·mol⁻¹ | C32H31N5O2 | 25 MG
SDP
Catalog No. 5000245428
Encompass_Preferred
P-gp inhibitor 1 is a small-molecule inhibitor that reverses P-glycoprotein (ABCB1/MDR1)-mediated multidrug resistance. Supplied as a purified solid for research use, it is intended for use in in vitro transporter inhibition assays, drug-drug interaction studies, and cellular models of chemoresistance. Not for human or veterinary use.
  • Reverses P-glycoprotein-mediated multidrug resistance in cellular assays.
  • High purity suitable for biochemical and cellular studies.
  • Soluble in common organic solvents for assay preparation.
  • Useful for transporter inhibition and drug-interaction profiling.
  • Supplied as a measured mass for reproducible dosing in experiments.
  • Intended for research use only.
Catalog No. 50-002-45428 Supplier Medchemexpress LLC Supplier No. HY10179125MG
P-gp inhibitor 1 is a small-molecule inhibitor that reverses P-glycoprotein (ABCB1/MDR1)-mediated multidrug resistance. Supplied as a purified solid for research use, it is intended for use in in vitro transporter inhibition assays, drug-drug interaction studies, and cellular models of chemoresistance. Not for human or veterinary use.
  • Reverses P-glycoprotein-mediated multidrug resistance in cellular assays.
  • High purity suitable for biochemical and cellular studies.
  • Soluble in common organic solvents for assay preparation.
  • Useful for transporter inhibition and drug-interaction profiling.
  • Supplied as a measured mass for reproducible dosing in experiments.
  • Intended for research use only.