Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC P-gp inhibitor 1 | 2050747-49-2 | 98.8% | 517.62 g/mol | C32H31N5O2 | 5 MG
SDP

Catalog No. 5000245578
Encompass_Preferred

P-gp inhibitor 1 is a small-molecule research reagent that inhibits P-glycoprotein and can reverse P-glycoprotein-mediated multidrug resistance in vitro. It is supplied as a light yellow to brown solid of high purity for laboratory research use and is accompanied by analytical data and handling documents.

  • High purity solid suitable for in vitro studies.
  • Soluble in DMSO (approx. 16.67 mg/mL with warming).
  • Supplied with analytical documents: COA, HNMR, LC-MS.
  • Storage recommendations for stability provided.
  • Intended for research use only, not for human use.

Catalog No. 50-002-45578 Supplier Medchemexpress LLC Supplier No. HY1017915MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

P-gp inhibitor 1 is a small-molecule research reagent that inhibits P-glycoprotein and can reverse P-glycoprotein-mediated multidrug resistance in vitro. It is supplied as a light yellow to brown solid of high purity for laboratory research use and is accompanied by analytical data and handling documents.

  • High purity solid suitable for in vitro studies.
  • Soluble in DMSO (approx. 16.67 mg/mL with warming).
  • Supplied with analytical documents: COA, HNMR, LC-MS.
  • Storage recommendations for stability provided.
  • Intended for research use only, not for human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.