Learn More
Grainger MASTER APPLIANCE Heat Gun, Pistol Grip
Catalog No. NC2326801
Encompass_Preferred
The Master Appliance Heat Gun is a 2000-watt pistol grip heat gun that provides powerful, concentrated heat for stripping paint, thawing pipes, bending plastics, shrinking tubing, and other heat applications. It features two heat settings (750°F and 1000°F), a 6 foot cord, and a lightweight, ergonomic composite housing for user comfort and control. The heat gun has overload protection and comes with two nozzles for directing heat flow.
Catalog No. NC2326801 Supplier Grainger Supplier No. 54XY07
The Master Appliance Heat Gun is a 2000-watt pistol grip heat gun that provides powerful, concentrated heat for stripping paint, thawing pipes, bending plastics, shrinking tubing, and other heat applications. It features two heat settings (750°F and 1000°F), a 6 foot cord, and a lightweight, ergonomic composite housing for user comfort and control. The heat gun has overload protection and comes with two nozzles for directing heat flow.