Learn More
ABclonal Technology p-GSK3B-Y216 - GSK3a-Y279
SDP
Supplier:  ABclonal Technology AP1530100UL
Encompass_Preferred
The protein encoded by this gene is a serine-threonine kinase belonging to the glycogen synthase kinase subfamily It is involved in energy metabolism neuronal cell development and body pattern formation Polymorphisms in this gene have been implicated in modifying risk of Parkinson disease and studies in mice show that overexpression of this gene may be relevant to the pathogenesis of Alzheimer disease Alternatively spliced transcript variants encoding different isoforms have been found for this gene
Catalog No. 50-000-13663