Learn More
CPC Scientific H-Pro-His-Pro-Phe-His-Phe-Phe-Val-Tyr-Lys-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific RENI010A
Encompass
Specific and at physiological pH soluable inhibitor of human renin in vitro.ONE-LETTER SEQUENCE: PHPFHFFVYKMOLECULAR FORMULA: C69H87N15O12MOLECULAR WEIGHT:1318.6STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [75645-19-1], RESEARCH AREA: CardiovascularREFERENCES: Burton, J., et al., Proc. Nat. Acad. Sci. USA, 77, 5476 (1980)
Catalog No. 50-211-0366