Learn More
Lonza Walkersville P-HDF - Diseased - Lesion
Catalog No. 5000496264
Encompass
Psoriasis is believed to be an immune related condition characterized by patches of abnormal skin Psoriasis is chronic lasting years or even an entire lifetime and impacts millions of individuals every year Psoriasis is believed to have a genetic component however scientists are still uncertain what specifically causes the disease For individuals with psoriasis skin cells grow at an abnormal rate causing the build-up of lesion sites These flair-ups are often initiated by a trigger such as stress or injury Lonza offers dermal fibroblasts isolated from both lesion sites and non-lesion sites all isolated from the same donor diagnosed with psoriasis Catalog no 219736 (Lot no 6F5008)Donor Information 69 year old Caucasian maleDisease Psoriasis (Lesion Site Derived)Culture Media Lonza FGM-2 Fibroblast Growth Medium BulletKit (Lonza Cat No
Catalog No. 50-004-96264 Supplier Lonza Walkersville Supplier No. 00219736
Psoriasis is believed to be an immune related condition characterized by patches of abnormal skin Psoriasis is chronic lasting years or even an entire lifetime and impacts millions of individuals every year Psoriasis is believed to have a genetic component however scientists are still uncertain what specifically causes the disease For individuals with psoriasis skin cells grow at an abnormal rate causing the build-up of lesion sites These flair-ups are often initiated by a trigger such as stress or injury Lonza offers dermal fibroblasts isolated from both lesion sites and non-lesion sites all isolated from the same donor diagnosed with psoriasis Catalog no 219736 (Lot no 6F5008)Donor Information 69 year old Caucasian maleDisease Psoriasis (Lesion Site Derived)Culture Media Lonza FGM-2 Fibroblast Growth Medium BulletKit (Lonza Cat No