Learn More
Selleck Chemical LLC p-Hydroxybenzaldehyde
SDP
Catalog No. 5000002998
Encompass_Preferred
p-Hydroxybenzaldehyde (4-Hydroxybenzaldehyde 4-Formylphenol p-Formylphenol) which can be found in the orchids Gastrodia elata Galeola faberi and vanilla is a hydroxybenzaldehyde that reacts with NAD and H2O to produce 4-hydroxybenzoate NADH and 2 protons p-Hydroxybenzaldehyde at 101 7 M can significantly reduce the GABA-induced chloride current of GABAA receptors( 1 2 2S subtype) expressed
Catalog No. 50-000-02998 Supplier Selleck Chemical LLC Supplier No. S600825MG
p-Hydroxybenzaldehyde (4-Hydroxybenzaldehyde 4-Formylphenol p-Formylphenol) which can be found in the orchids Gastrodia elata Galeola faberi and vanilla is a hydroxybenzaldehyde that reacts with NAD and H2O to produce 4-hydroxybenzoate NADH and 2 protons p-Hydroxybenzaldehyde at 101 7 M can significantly reduce the GABA-induced chloride current of GABAA receptors( 1 2 2S subtype) expressed