Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

P-Hydroxybenzoic Acid Crystalline MP Biomedicals

Catalog No. ICN10257780 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10257780 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10257780 Supplier MP Biomedicals, Inc Supplier No. 0210257780
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 99-96-7
Molecular Formula C7H6O3
Molecular Weight (g/mol) 138.12
MDL Number MFCD00002547
InChI Key FJKROLUGYXJWQN-UHFFFAOYSA-N
Synonym p-hydroxybenzoic acid, 4-carboxyphenol, benzoic acid, 4-hydroxy, p-salicylic acid, benzoic acid, p-hydroxy, para-hydroxybenzoic acid, 4-hydroxybenzoesaeure, p-carboxyphenol, 4-hydroxy benzoic acid, p-oxybenzoesaure german
PubChem CID 135
ChEBI CHEBI:30763
IUPAC Name 4-hydroxybenzoic acid
SMILES OC(=O)C1=CC=C(O)C=C1

Specifications

Density 1.46 g/mL
Quantity 100 g
Formula Weight 138.1
Physical Form Crystalline
Chemical Name or Material P-Hydroxybenzoic Acid
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.