Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC p-Hydroxycinnamic acid | 7400-08-0 | 10 G
SDP

Catalog No. 5000420344
Encompass_Preferred

p-Hydroxycinnamic acid is a chemical compound (CAS No. 7400-08-0) primarily utilized as a laboratory chemical and in the manufacture of other substances. With a molecular formula of C9H8O3 and a molecular weight of 164.16, this white to off-white solid is stable under recommended storage conditions and is intended for research use only.

  • Appears as a white to off-white solid
  • Purity of 99.94% by HPLC
  • Structure consistent with ¹H NMR spectrum
  • Stable under recommended storage conditions
  • Melting point ranges from 212-214°C
  • Store powder at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 2 years or -20°C for 1 year
  • Requires careful handling to avoid skin, eye, and respiratory irritation

Catalog No. 50-004-20344 Supplier Medchemexpress LLC Supplier No. HYN239110G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

p-Hydroxycinnamic acid is a chemical compound (CAS No. 7400-08-0) primarily utilized as a laboratory chemical and in the manufacture of other substances. With a molecular formula of C9H8O3 and a molecular weight of 164.16, this white to off-white solid is stable under recommended storage conditions and is intended for research use only.

  • Appears as a white to off-white solid
  • Purity of 99.94% by HPLC
  • Structure consistent with ¹H NMR spectrum
  • Stable under recommended storage conditions
  • Melting point ranges from 212-214°C
  • Store powder at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 2 years or -20°C for 1 year
  • Requires careful handling to avoid skin, eye, and respiratory irritation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.