Learn More
Medchemexpress LLC P-Hydroxycinnamic acid | 7400-08-0 | 100 G
SDP
Catalog No. 5000420679
Encompass_Preferred
p-Hydroxycinnamic acid is a common dietary phenol and ubiquitous plant metabolite that can inhibit platelet activity, thromboxane B2 production, and lipopolysaccharide-induced prostaglandin E2 generation. It also exhibits antioxidant and anti-inflammatory properties.
  • Inhibits platelet activity.
  • Inhibits thromboxane B2 production (IC50 of 371 μM).
  • Inhibits lipopolysaccharide-induced prostaglandin E2 generation (IC50 of 126 μM).
  • Possesses antioxidant properties.
  • Possesses anti-inflammatory properties.
  • Reduces ADP-induced platelet aggregation.
  • Interferes with the arachidonic acid cascade.
  • Has a purity of 99.94%.
Catalog No. 50-004-20679 Supplier Medchemexpress LLC Supplier No. HYN2391100G
p-Hydroxycinnamic acid is a common dietary phenol and ubiquitous plant metabolite that can inhibit platelet activity, thromboxane B2 production, and lipopolysaccharide-induced prostaglandin E2 generation. It also exhibits antioxidant and anti-inflammatory properties.
  • Inhibits platelet activity.
  • Inhibits thromboxane B2 production (IC50 of 371 μM).
  • Inhibits lipopolysaccharide-induced prostaglandin E2 generation (IC50 of 126 μM).
  • Possesses antioxidant properties.
  • Possesses anti-inflammatory properties.
  • Reduces ADP-induced platelet aggregation.
  • Interferes with the arachidonic acid cascade.
  • Has a purity of 99.94%.