Learn More
Medchemexpress LLC p-Hydroxycinnamic acid | 7400-08-0 | 50 G
SDP
Catalog No. 5000420325
Encompass_Preferred
p-Hydroxycinnamic acid is a common dietary phenol that inhibits platelet activity and interferes with the arachidonic acid cascade. It reduces ADP-induced platelet aggregation and affects thromboxane B2 production and prostaglandin E2 generation.
  • Inhibits platelet activity
  • Reduces ADP-induced platelet aggregation
  • Interferes with the arachidonic acid cascade
  • Reduces thromboxane B2 production
  • Reduces lipopolysaccharide-induced prostaglandin E2 generation
Catalog No. 50-004-20325 Supplier Medchemexpress LLC Supplier No. HYN239150G
p-Hydroxycinnamic acid is a common dietary phenol that inhibits platelet activity and interferes with the arachidonic acid cascade. It reduces ADP-induced platelet aggregation and affects thromboxane B2 production and prostaglandin E2 generation.
  • Inhibits platelet activity
  • Reduces ADP-induced platelet aggregation
  • Interferes with the arachidonic acid cascade
  • Reduces thromboxane B2 production
  • Reduces lipopolysaccharide-induced prostaglandin E2 generation