Learn More
Medchemexpress LLC p-Hydroxymandelic acid | 1198-84-1 | 95.5% | 168.15 | 5 G
SDP
Catalog No. 5000439606
Encompass_Preferred
p-Hydroxymandelic acid is a valuable aromatic fine chemical used extensively in the production of pharmaceuticals and food additives. It serves as an important precursor for synthesizing 4-hydroxyphenylacetic acid, which is then used to create atenolol, a selective β1-receptor antagonist drug. This compound has also been utilized in adaptor systems to conjugate cytotoxic drugs and enzyme substrates, showing potential for targeted drug delivery systems.
  • Used in the production of aromatic drugs and flavors.
  • Precursor for 4-hydroxyphenylacetic acid, used in the synthesis of atenolol.
  • Can conjugate cytotoxic drugs and enzyme substrates for targeted drug delivery systems.
Catalog No. 50-004-39606 Supplier Medchemexpress LLC Supplier No. HY1130275G
Add to Cart
Add to Cart
p-Hydroxymandelic acid is a valuable aromatic fine chemical used extensively in the production of pharmaceuticals and food additives. It serves as an important precursor for synthesizing 4-hydroxyphenylacetic acid, which is then used to create atenolol, a selective β1-receptor antagonist drug. This compound has also been utilized in adaptor systems to conjugate cytotoxic drugs and enzyme substrates, showing potential for targeted drug delivery systems.
  • Used in the production of aromatic drugs and flavors.
  • Precursor for 4-hydroxyphenylacetic acid, used in the synthesis of atenolol.
  • Can conjugate cytotoxic drugs and enzyme substrates for targeted drug delivery systems.