Learn More
Medchemexpress LLC p-Hydroxymandelic acid | 1198-84-1 | 95.5% | 168.15 | 500 MG
SDP
Catalog No. 5000439416
Encompass_Preferred
p-Hydroxymandelic acid is a valuable aromatic fine chemical and widely used for production of pharmaceuticals and food additives. It is also used in the production of aromatic drugs and flavors, and for the preparation of 4-hydroxyphenylacetic acid, a synthetic precursor of the selective β1-receptor antagonist drug atenolol. Additionally, p-Hydroxymandelic acid can conjugate cytotoxic drugs and enzyme substrates, suggesting its application in targeted drug delivery systems.
  • Valuable aromatic fine chemical.
  • Used for production of pharmaceuticals.
  • Used for production of food additives.
  • Applied in targeted drug delivery systems.
Catalog No. 50-004-39416 Supplier Medchemexpress LLC Supplier No. HY113027500MG
p-Hydroxymandelic acid is a valuable aromatic fine chemical and widely used for production of pharmaceuticals and food additives. It is also used in the production of aromatic drugs and flavors, and for the preparation of 4-hydroxyphenylacetic acid, a synthetic precursor of the selective β1-receptor antagonist drug atenolol. Additionally, p-Hydroxymandelic acid can conjugate cytotoxic drugs and enzyme substrates, suggesting its application in targeted drug delivery systems.
  • Valuable aromatic fine chemical.
  • Used for production of pharmaceuticals.
  • Used for production of food additives.
  • Applied in targeted drug delivery systems.