Learn More
Medchemexpress LLC p-Hydroxymandelic acid | 1198-84-1 | 98.9% | 168.15 | 25 G
SDP
Catalog No. 5000439522
Encompass_Preferred
p-Hydroxymandelic acid is a valuable aromatic fine chemical widely used for the production of pharmaceuticals and food additives. It is also known as 4-Hydroxymandelic acid (4-HMA) and serves as a synthetic precursor for 4-hydroxyphenylacetic acid, a key intermediate in the synthesis of the β1-receptor antagonist drug atenolol.
  • Valuable aromatic fine chemical
  • Used for production of pharmaceuticals
  • Used for production of food additives
  • Employed for preparation of 4-hydroxyphenylacetic acid
  • Can conjugate cytotoxic drugs and enzyme substrates
  • Promising application in targeted drug delivery systems
Catalog No. 50-004-39522 Supplier Medchemexpress LLC Supplier No. 25GHY113027
p-Hydroxymandelic acid is a valuable aromatic fine chemical widely used for the production of pharmaceuticals and food additives. It is also known as 4-Hydroxymandelic acid (4-HMA) and serves as a synthetic precursor for 4-hydroxyphenylacetic acid, a key intermediate in the synthesis of the β1-receptor antagonist drug atenolol.
  • Valuable aromatic fine chemical
  • Used for production of pharmaceuticals
  • Used for production of food additives
  • Employed for preparation of 4-hydroxyphenylacetic acid
  • Can conjugate cytotoxic drugs and enzyme substrates
  • Promising application in targeted drug delivery systems