Learn More
TARGETMOL CHEMICALS INC P-Hydroxyphenethyl trans-ferul
SDP
Catalog No. 502880281
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. P-Hydroxyphenethyl trans-ferulate exhibits affinity toward 5-HT(7) receptors in a competitive binding assay. P-Hydroxyphenethyl trans-ferulate can double quinone reductase specific activity in Hepa 1c1c7 cells at a level of 2.1 microg/mL (6.6 microM). Purity 99.3%
Catalog No. 50-288-0281 Supplier TARGETMOL CHEMICALS INC Supplier No. TN20305MG
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. P-Hydroxyphenethyl trans-ferulate exhibits affinity toward 5-HT(7) receptors in a competitive binding assay. P-Hydroxyphenethyl trans-ferulate can double quinone reductase specific activity in Hepa 1c1c7 cells at a level of 2.1 microg/mL (6.6 microM). Purity 99.3%