Learn More
Medchemexpress LLC p-Hydroxyphenethyl trans-ferulate | 84873-15-4 | 5 MG
SDP
Catalog No. 5000420631
Encompass_Preferred
p-Hydroxyphenethyl trans-ferulate is a compound known for its diverse biological activities. It exhibits anti-hyperglycemic, antioxidant, and anti-inflammatory properties, alongside inhibitory anticancer and serotonergic activities.
  • Exhibits anti-hyperglycemic activity against yeast α-glucosidase (IC50=19.24 ± 1.73 μM)
  • Possesses antioxidant properties
  • Demonstrates anti-inflammatory activities
  • Shows inhibitory anticancer activity against various human cell lines
  • Exhibits serotonergic activity by displacing [3H]LSD from human recombinant 5HT7 receptor
  • Has anticomplement activity in sheep and rabbit erythrocytes
  • High purity of 99.68%
Catalog No. 50-004-20631 Supplier Medchemexpress LLC Supplier No. HYN30785MG
p-Hydroxyphenethyl trans-ferulate is a compound known for its diverse biological activities. It exhibits anti-hyperglycemic, antioxidant, and anti-inflammatory properties, alongside inhibitory anticancer and serotonergic activities.
  • Exhibits anti-hyperglycemic activity against yeast α-glucosidase (IC50=19.24 ± 1.73 μM)
  • Possesses antioxidant properties
  • Demonstrates anti-inflammatory activities
  • Shows inhibitory anticancer activity against various human cell lines
  • Exhibits serotonergic activity by displacing [3H]LSD from human recombinant 5HT7 receptor
  • Has anticomplement activity in sheep and rabbit erythrocytes
  • High purity of 99.68%