Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Iodobenzoic Acid, MP Biomedicals

Catalog No. ICN213202 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN213202 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN213202 Supplier MP Biomedicals, Inc Supplier No. 0521320205

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 619-58-9
Molecular Formula C7H5IO2
Molecular Weight (g/mol) 248.02
MDL Number MFCD00002533
InChI Key GHICCUXQJBDNRN-UHFFFAOYSA-N
Synonym p-iodobenzoic acid, benzoic acid, 4-iodo, benzoic acid, p-iodo, 4-iodo-benzoic acid, p-iodobenzenecarboxylic acid, 4-jodbenzoesaeure, unii-ipo4lyq1en, 4-jodbenzoesaeure german, 4-iodo benzoic acid, ipo4lyq1en
PubChem CID 12085
IUPAC Name 4-iodobenzoic acid
SMILES OC(=O)C1=CC=C(I)C=C1

Specifications

Quantity 10 g
Formula Weight 248.02
Chemical Name or Material p-Iodobenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.