Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Iodoclonidine hydrochloride, MilliporeSigma™ Supelco™

Catalog No. 111012428 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2428 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-2428 Supplier MilliporeSigma Supelco Supplier No. I11425MGI
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 108294-53-7
Physical Form Solid
Linear Formula C9H8 Cl2 IN3 · HCl
MDL Number MFCD00069292
Synonym 2-[(2,6-Dichloro-4-iodophenyl)imino]imidazoline hydrochloride
Solubility Soluble in water
Type Analytical Standard
Industry Type Drug Analysis
Quantity 25 mg
Formula Weight 392.45
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.