Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Iodonitrotetazolium violet, 100.7%, MP Biomedicals™

Catalog No. ICN10206601 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10206601 1 g
ICN10206605 5 g
ICN10206625 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN10206601 Supplier MP Biomedicals, Inc Supplier No. 0210206601
Add to Cart
Add to Cart

Chemical Identifiers

CAS 146-68-9
Molecular Formula C19H13ClIN5O2
Molecular Weight (g/mol) 505.70
MDL Number MFCD00011961,MFCD00149999
InChI Key JORABGDXCIBAFL-UHFFFAOYSA-M
Synonym iodonitrotetrazolium chloride, iodonitrotetrazolium, iodonitrotetrazolium purple, iodonitrotetrazolium violet, 2-4-iodophenyl-3-4-nitrophenyl-5-phenyltetrazolium chloride, iodonitro tetrazolium, int, p-iodonitrotetrazolium violet, int chemical, 2-p-iodophenyl-3-p-nitrophenyl-5-phenyltetrazolium chloride
PubChem CID 64957
ChEBI CHEBI:75421
IUPAC Name 3-(4-iodophenyl)-2-(4-nitrophenyl)-5-phenyl-3H-1,2λ⁵,3,4-tetrazol-2-ylium chloride
SMILES [Cl-].[O-][N+](=O)C1=CC=C(C=C1)[N+]1=NC(=NN1C1=CC=C(I)C=C1)C1=CC=CC=C1

Specifications

Color Yellow
Quantity 1 g
Formula Weight 505.7
Percent Purity 1.007
Physical Form Fine Powder
Chemical Name or Material p-Iodonitrotetazolium violet
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.