Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Iodonitrotetrazolium Violet, MP Biomedicals

Catalog No. MP021020665 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP021020665 500 mg
MP021020661 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP021020665 Supplier MP Biomedicals, Inc Supplier No. 02102066.5
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

  • p-Iodonitrotetazolium Violet is used as a electron acceptor for the colorimetric assay of various dehydrogenases

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 146-68-9
Molecular Formula C19H13ClIN5O2
Molecular Weight (g/mol) 505.70
MDL Number MFCD00011961,MFCD00149999
InChI Key JORABGDXCIBAFL-UHFFFAOYSA-M
Synonym iodonitrotetrazolium chloride, iodonitrotetrazolium, iodonitrotetrazolium purple, iodonitrotetrazolium violet, 2-4-iodophenyl-3-4-nitrophenyl-5-phenyltetrazolium chloride, iodonitro tetrazolium, int, p-iodonitrotetrazolium violet, int chemical, 2-p-iodophenyl-3-p-nitrophenyl-5-phenyltetrazolium chloride
PubChem CID 64957
ChEBI CHEBI:75421
IUPAC Name 3-(4-iodophenyl)-2-(4-nitrophenyl)-5-phenyl-3H-1,2λ⁵,3,4-tetrazol-2-ylium chloride
SMILES [Cl-].[O-][N+](=O)C1=CC=C(C=C1)[N+]1=NC(=NN1C1=CC=C(I)C=C1)C1=CC=CC=C1

Specifications

Melting Point 245°C to 251°C (literature)
Color Yellow
Linear Formula C19H13ClIN5O2
Quantity 500 mg
Solubility Information Soluble in 5% methanol/water(1:1)(clear yellow); slightly soluble in water, alcohol, DMSO.
Formula Weight 505.7
Physical Form Powder
Chemical Name or Material p-IODONITROTETRAZOLIUM VIOLET
Recommended Storage Store at +4°C, desiccate, protect from light.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.