Learn More
Thermo Scientific P-Length Tubing 1 Roll
Supplier:  Thermo Scientific CH106508
Encompass
Plastic PTFE, FEP, or PFA tubing for short-term fluid transfer in laboratories, with smooth inner wall for low binding and high flow, withstands temperatures up to 200°C, requires push-to-connect or compression fittings for leak-free connections.
Catalog No. NC9941640