Learn More
Strem, An Ascensus Company CAS# 2179272-79-6. 100mg. 1-[2,6-Bis(cyclohexyloxy)phenyl]-2-(di-tertbutylphosphaneyl)-1H-imidazole, min. 95%.
SDP
Supplier:  Strem, An Ascensus Company 153360100MG
Encompass
CAS# 2179272-79-6. 100mg. 1-[2,6-Bis(cyclohexyloxy)phenyl]-2-(di-tertbutylphosphaneyl)-1H-imidazole, min. 95%. . Molecular Weight: 484.66. Molecular Formula: C29H45N2O2P. Color/form: white to off-white to pale yellow solid Strem# 15-3360. http://www.strem.com/catalog/v/15-3360/
Catalog No. 50-206-5426