Learn More
TARGETMOL CHEMICALS INC p-MPPF 1MG
SDP
Catalog No. 502877865
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. p-MPPF is a selective 5-HT antagonist that dose-dependently antagonizes hypothermia induced by 8-hydroxy-2-(di-n-propylamino)tetralin (8-OH-DPAT) and can be used to study neurological diseases. Purity 99.79%
Catalog No. 50-287-7865 Supplier TARGETMOL CHEMICALS INC Supplier No. T69750L1MG
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. p-MPPF is a selective 5-HT antagonist that dose-dependently antagonizes hypothermia induced by 8-hydroxy-2-(di-n-propylamino)tetralin (8-OH-DPAT) and can be used to study neurological diseases. Purity 99.79%