Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC p-MPPI hydrochloride | 220643-77-6 | 99.8% | 25 MG
SDP

Catalog No. 5000234836
Encompass_Preferred

p-MPPI hydrochloride is a research-grade small molecule that acts as a selective 5-HT1A receptor antagonist and is supplied as the hydrochloride salt for pharmacological and biochemical studies.

  • Selective 5-HT1A receptor antagonist for receptor pharmacology studies.
  • High purity suitable for research use (reported 99.8%).
  • Molecular weight 578.87 g/mol; formula C25H28ClIN4O2.
  • Soluble in DMSO and water for in vitro applications.
  • Available in small research quantities, such as 25 mg.

Catalog No. 50-002-34836 Supplier Medchemexpress LLC Supplier No. HY12073825MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

p-MPPI hydrochloride is a research-grade small molecule that acts as a selective 5-HT1A receptor antagonist and is supplied as the hydrochloride salt for pharmacological and biochemical studies.

  • Selective 5-HT1A receptor antagonist for receptor pharmacology studies.
  • High purity suitable for research use (reported 99.8%).
  • Molecular weight 578.87 g/mol; formula C25H28ClIN4O2.
  • Soluble in DMSO and water for in vitro applications.
  • Available in small research quantities, such as 25 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.