Learn More
p-Naphtholbenzein Indicator, 1% in Titration Solvent, Reagents
Specifications
Specifications
| Chemical Name or Material | p-Naphtholbenzein Indicator, 1% in Titration Solvent ASTM Methods D974, D3242, D3339 |
| Boiling Point | 90°C |
| CAS | 108-88-3, 67-63-0, 7732-18-5 |
| Synonym | alpha-Naphtholbenzein, para-Naphtholbenzein, 1-Naphtholbenzein |
| Grade | ASTM |
| Packaging | Glass Bottle |
| Specific Gravity | 0.8245 |
| Color | Dark Red |
| Quantity | 1 L |
| Physical Form | Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.