Learn More
Aqua Solutions p-Naptholbenzein 1% w/v Indicator Solution (Size: 1L)
SDP
Catalog No. NC3684682
Encompass_Preferred
Prepared to ASTM D3242 and D974, , , DOT: Flammable liquid, n.o.s. (Toluene, Isopropanol), 3, UN1993, PG II
Catalog No. NC3684682 Supplier Aqua Solutions Supplier No. 61001L
Add to Cart
Add to Cart
Prepared to ASTM D3242 and D974, , , DOT: Flammable liquid, n.o.s. (Toluene, Isopropanol), 3, UN1993, PG II