Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC p-NCS-benzyl-NODA-GA | 1660127-21-8 | 99.5% | 521.59 g/mol | C23H31N5O7S | 10 MG
SDP

Catalog No. 5000288244
Encompass_Preferred

p-NCS-benzyl-NODA-GA is a bifunctional chelator that enables covalent conjugation to biomolecules and stable metal ion complexation for imaging and radiolabeling. It contains a para-isothiocyanatobenzyl reactive group for attachment to primary amines and is supplied for research use only.

  • Bifunctional chelator for bioconjugation and radiolabeling.
  • Isothiocyanate functional group reacts with primary amines.
  • High purity (>99%) suitable for analytical and preparative work.
  • Molecular weight 521.59 g/mol; formula C23H31N5O7S.
  • Soluble in DMSO (50 mg/mL) with ultrasonic assistance.
  • Available as small solid quantities and as 10 mM solution in DMSO.
  • Stored cold in solvent: -80°C (6 months) or -20°C (1 month) in sealed, dry conditions.

Catalog No. 50-002-88244 Supplier Medchemexpress LLC Supplier No. HY15918010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

p-NCS-benzyl-NODA-GA is a bifunctional chelator that enables covalent conjugation to biomolecules and stable metal ion complexation for imaging and radiolabeling. It contains a para-isothiocyanatobenzyl reactive group for attachment to primary amines and is supplied for research use only.

  • Bifunctional chelator for bioconjugation and radiolabeling.
  • Isothiocyanate functional group reacts with primary amines.
  • High purity (>99%) suitable for analytical and preparative work.
  • Molecular weight 521.59 g/mol; formula C23H31N5O7S.
  • Soluble in DMSO (50 mg/mL) with ultrasonic assistance.
  • Available as small solid quantities and as 10 mM solution in DMSO.
  • Stored cold in solvent: -80°C (6 months) or -20°C (1 month) in sealed, dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.