Learn More
Medchemexpress LLC P-NH2-Bn-NOTA hydrochloride hydrate | 142131-37-1 | 98.1% | 517.4 g/mol | C19H34Cl2N4O8 | 10 MG
SDP
Catalog No. 5000288409
Encompass_Preferred
p-NH2-Bn-NOTA hydrochloride hydrate is a bifunctional NOTA-derived chelator supplied as the hydrochloride hydrate salt for use in bioconjugation and radiolabeling applications. It features an amine-functionalized benzyl linker that facilitates coupling to peptides, antibodies, or other biomolecules for preclinical imaging and tumor-targeting studies.
  • Bifunctional macrocyclic NOTA chelator suitable for radiometal complexation.
  • Amine-functionalized benzyl linker enables straightforward conjugation to biomolecules.
  • Provided as hydrochloride hydrate for improved handling and stability.
  • Available in small milligram pack sizes to support research-scale use.
  • Accompanied by COA and SDS documents for quality and safety information.
Catalog No. 50-002-88409 Supplier Medchemexpress LLC Supplier No. HY15806610MG
p-NH2-Bn-NOTA hydrochloride hydrate is a bifunctional NOTA-derived chelator supplied as the hydrochloride hydrate salt for use in bioconjugation and radiolabeling applications. It features an amine-functionalized benzyl linker that facilitates coupling to peptides, antibodies, or other biomolecules for preclinical imaging and tumor-targeting studies.
  • Bifunctional macrocyclic NOTA chelator suitable for radiometal complexation.
  • Amine-functionalized benzyl linker enables straightforward conjugation to biomolecules.
  • Provided as hydrochloride hydrate for improved handling and stability.
  • Available in small milligram pack sizes to support research-scale use.
  • Accompanied by COA and SDS documents for quality and safety information.