Learn More
Cayman Chemical p-nItro-PIfIthrIn-a 10mg
SDP
Catalog No. 502970211
Encompass_Preferred
A cell-permeable analog of pifithrin-α that blocks p53-mediated expression of p21/WAF1 and apoptosis in cortical neurons ten-fold more potently than pifithrin-α; suppresses p53-mediated TGF-β1 expression in human proximal tubular cells and attenuates steatosis and liver injury in mice fed a high-fat diet
Catalog No. 50-297-0211 Supplier Cayman Chemical Supplier No. 1620910 MG
A cell-permeable analog of pifithrin-α that blocks p53-mediated expression of p21/WAF1 and apoptosis in cortical neurons ten-fold more potently than pifithrin-α; suppresses p53-mediated TGF-β1 expression in human proximal tubular cells and attenuates steatosis and liver injury in mice fed a high-fat diet