Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Nitroaniline, 98.9%, MP Biomedicals™

Catalog No. ICN19025380 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
ICN19025380 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19025380 Supplier MP Biomedicals, Inc Supplier No. 0219025380

Specifications

CAS 100-01-6
Color Yellow
Density 1.44g/cm3 at 20°C
Assay Percent Range 0.989
Molecular Formula C6H6N2O2
Quantity 100 g
MDL Number MFCD00007858
Synonym p-nitroaniline, p-nitraniline, 4-nitrobenzenamine, p-aminonitrobenzene, 4-nitraniline, benzenamine, 4-nitro, p-nitrophenylamine, 1-amino-4-nitrobenzene, aniline, p-nitro, developer p
InChI Key TYMLOMAKGOJONV-UHFFFAOYSA-N
SMILES NC1=CC=C(C=C1)[N+]([O-])=O
IUPAC Name 4-nitroaniline
Molecular Weight (g/mol) 138.13
PubChem CID 7475
ChEBI CHEBI:17064
Formula Weight 138.12
Physical Form Crystalline Powder
Chemical Name or Material p-Nitroaniline
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.