Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Nitrophenol, >99%, MP Biomedicals™

Catalog No. ICN10246125 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10246125 25 g
ICN10246101 1 g
ICN10246180 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN10246125 Supplier MP Biomedicals, Inc Supplier No. 0210246125
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 100-02-7
Molecular Formula C6H5NO3
Molecular Weight (g/mol) 139.11
MDL Number MFCD00007331
InChI Key BTJIUGUIPKRLHP-UHFFFAOYSA-N
Synonym p-nitrophenol, phenol, 4-nitro, paranitrophenol, niphen, 4-hydroxynitrobenzene, p-hydroxynitrobenzene, phenol, p-nitro, mononitrophenol, paranitrofenol, paranitrofenolo
PubChem CID 980
ChEBI CHEBI:16836
IUPAC Name 4-nitrophenol
SMILES OC1=CC=C(C=C1)[N+]([O-])=O

Specifications

Color Yellow
Density 1.479g/mL
Quantity 25 g
Formula Weight 139.11
Percent Purity >99%
Physical Form Crystalline Powder
Chemical Name or Material p-Nitrophenol
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.