Learn More
Crescent Chemical Co Inc P-NITROPHENOL SOLN 1ML
SDP
Catalog No. 5068940
Encompass_Preferred
p-Nitrophenol Single Component Calibration Standard in Dichloromethane at 1000 ug/mL 1mL
Catalog No. 50-689-40 Supplier Crescent Chemical Co Inc Supplier No. CT3301D.10
p-Nitrophenol Single Component Calibration Standard in Dichloromethane at 1000 ug/mL 1mL