Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Nitrophenyl Acetate, MP Biomedicals™

Catalog No. ICN10088805 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10088805 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10088805 Supplier MP Biomedicals, Inc Supplier No. 0210088805
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Synthetic substrate used in assays for esterase and lipase activity.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 830-03-5
Molecular Formula C8H7NO4
Molecular Weight (g/mol) 181.15
MDL Number MFCD00007326
InChI Key QAUUDNIGJSLPSX-UHFFFAOYSA-N
Synonym p-nitrophenyl acetate, p-acetoxynitrobenzene, acetic acid, 4-nitrophenyl ester, p-nitrophenol acetate, acetic acid p-nitrophenyl ester, acetic acid, p-nitrophenyl ester, p-nitrophenyl acetate van, 4-nitrophenyl acetate, unii-i902j0qh9s, acetic acid 4-nitrophenyl ester
PubChem CID 13243
ChEBI CHEBI:82635
IUPAC Name 4-nitrophenyl acetate
SMILES CC(=O)OC1=CC=C(C=C1)[N+]([O-])=O

Specifications

Color White to Yellow
Quantity 5 g
Formula Weight 181.1
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material p-Nitrophenyl acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.