Learn More
Research Products International Corp p-Nitrophenyl-B-D-galactopyranoside, 1 G
SDP
Catalog No. 501978127
Encompass_Preferred
Colorimetric and spectrophotoametric substrate for detection of B-D Galactosidase.
Catalog No. 50-197-8127 Supplier Research Products International Corp Supplier No. N810101.0
Colorimetric and spectrophotoametric substrate for detection of B-D Galactosidase.